siRNA Details
Gene Symbol: Serp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030835.2 | NP_110462.1 | 3 | 1282 | ||
GenBank mRNA | AJ238236.1 | - | 3 | 942 | ||
BC061854.1 | - | 3 | 1282 | 5621665 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Serp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030835.2 | NP_110462.1 | 3 | 1282 | ||
GenBank mRNA | AJ238236.1 | - | 3 | 942 | ||
BC061854.1 | - | 3 | 1282 | 5621665 |