siRNA Details
| Gene Symbol: Erap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030836.1 | NP_110463.1 | 12, 13 | 1978 | ||
| XM_006231700.1 | XP_006231762.1 | 14, 15 | 2784 | |||
| XM_006231701.2 | XP_006231763.1 | 14, 15 | 2103 | |||
| XM_006231704.2 | XP_006231766.1 | 13, 14 | 2031 | |||
| XM_008760600.1 | XP_008758822.1 | 13, 14 | 4196 | |||
| XM_008760601.1 | XP_008758823.1 | 13, 14 | 4197 | |||
| XM_008760602.1 | XP_008758824.1 | 12, 13 | 4123 | |||
| GenBank mRNA | AF148323.1 | - | 12, 13 | 1975 | ||
| AF148324.1 | - | 12, 13 | 1978 | |||
| BC098664.1 | - | 13, 14 | 1985 | 7192921 | ||

Erap1
Genomic Map