siRNA Details
| Gene Symbol: Nptxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030841.3 | NP_110468.2 | 3 | 1182 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nptxr| Gene Symbol: Nptxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030841.3 | NP_110468.2 | 3 | 1182 | ||