siRNA Details
| Gene Symbol: Stxbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030843.2 | NP_110470.1 | 23 | 3271 | ||
| NM_178345.1 | NP_848035.1 | 25 | 3029 | |||
| NM_178346.1 | NP_848036.1 | 22 | 2870 | |||
| XM_006227623.2 | XP_006227685.1 | 26 | 3640 | |||
| XM_006227624.2 | XP_006227686.1 | 25 | 3592 | |||
| XM_008758628.1 | XP_008756850.1 | 24 | 3532 | |||
| XM_008758629.1 | XP_008756851.1 | 23 | 3481 | |||
| GenBank mRNA | AF118889.1 | - | 25 | 3029 | ||
| AF118890.1 | - | 22 | 2870 | |||
| FQ214558.1 | - | 7 | 1217 | |||
| U92072.1 | - | 23 | 3271 | |||

Stxbp5
Genomic Map