siRNA Details
| Gene Symbol: Bst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030848.1 | NP_110475.1 | 3 | 452 | ||
| GenBank mRNA | D49955.1 | - | 3 | 452 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bst1| Gene Symbol: Bst1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030848.1 | NP_110475.1 | 3 | 452 | ||
| GenBank mRNA | D49955.1 | - | 3 | 452 | ||