siRNA Details
| Gene Symbol: Bmpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030849.1 | NP_110476.1 | 11 | 1593 | ||
| GenBank mRNA | D17667.1 | - | 10 | 1368 | ||
| D38082.1 | - | 10 | 1450 | |||
| S75359.1 | - | 11 | 1593 | |||
| Gene Symbol: LOC686129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008767400.1 | XP_008765622.1 | 2 | 1613 | ||

Bmpr1a
Genomic Map