siRNA Details
| Gene Symbol: Bhmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030850.1 | NP_110477.1 | 6 | 855 | ||
| GenBank mRNA | AF038870.1 | - | 6 | 855 | ||
| BC078811.1 | - | 6 | 842 | 7130736 | ||
| FQ218356.1 | - | 1 | 178 | |||
| U96133.1 | - | 6 | 844 | |||

Bhmt
Genomic Map