siRNA Details
| Gene Symbol: Lat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030853.1 | NP_110480.1 | 3, 4 | 396 | ||
| XM_008759919.1 | XP_008758141.1 | 3, 4 | 850 | |||
| GenBank mRNA | AJ001184.1 | - | 3, 4 | 396 | ||
| BC088130.1 | - | 3, 4 | 356 | 7382238 | ||
| FQ233731.1 | - | 3, 4 | 413 | |||

Lat
Genomic Map