siRNA Details
| Gene Symbol: Lect1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030854.1 | NP_110481.1 | 3 | 460 | ||
| XM_006252346.1 | XP_006252408.1 | 3 | 455 | |||
| GenBank mRNA | AF051425.1 | - | 3 | 460 | ||
| BC127493.1 | - | 3 | 401 | 8369609 | ||

Lect1
Genomic Map