siRNA Details
| Gene Symbol: Smad7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030858.1 | NP_110485.1 | 4 | 2524 | ||
| XM_006254959.2 | XP_006255021.1 | 4 | 1118 | |||
| GenBank mRNA | AF042499.1 | - | 4 | 1246 | ||
| AF159626.1 | - | 4 | 2524 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smad7| Gene Symbol: Smad7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030858.1 | NP_110485.1 | 4 | 2524 | ||
| XM_006254959.2 | XP_006255021.1 | 4 | 1118 | |||
| GenBank mRNA | AF042499.1 | - | 4 | 1246 | ||
| AF159626.1 | - | 4 | 2524 | |||