siRNA Details
| Gene Symbol: Mtr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030864.1 | NP_110491.1 | 17, 18 | 2056 | ||
| GenBank mRNA | AF034214.1 | - | 17, 18 | 2056 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mtr| Gene Symbol: Mtr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030864.1 | NP_110491.1 | 17, 18 | 2056 | ||
| GenBank mRNA | AF034214.1 | - | 17, 18 | 2056 | ||