siRNA Details
| Gene Symbol: Ogg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030870.1 | NP_110497.1 | 2 | 222 | ||
| XM_008763198.1 | XP_008761420.1 | 2 | 434 | |||
| GenBank mRNA | AF029690.1 | - | 2 | 222 | ||
| BC166807.1 | - | 2 | 401 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ogg1| Gene Symbol: Ogg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030870.1 | NP_110497.1 | 2 | 222 | ||
| XM_008763198.1 | XP_008761420.1 | 2 | 434 | |||
| GenBank mRNA | AF029690.1 | - | 2 | 222 | ||
| BC166807.1 | - | 2 | 401 | |||