siRNA Details
| Gene Symbol: Pdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030872.1 | NP_110499.1 | 4 | 552 | ||
| GenBank mRNA | AF237719.1 | - | 4 | 552 | ||
| BC061823.1 | - | 4 | 516 | 5599188 | ||
| U10357.1 | - | 4 | 552 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pdk2| Gene Symbol: Pdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030872.1 | NP_110499.1 | 4 | 552 | ||
| GenBank mRNA | AF237719.1 | - | 4 | 552 | ||
| BC061823.1 | - | 4 | 516 | 5599188 | ||
| U10357.1 | - | 4 | 552 | |||