siRNA Details
| Gene Symbol: Pfn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030873.1 | NP_110500.1 | 232 | |||
| XM_006232390.2 | XP_006232452.1 | 2 | 357 | |||
| GenBank mRNA | AF228736.1 | - | 232 | |||
| AF228737.1 | - | 232 | ||||
| AY004289.1 | - | 232 | ||||
| BC087013.1 | - | 235 | ||||
| Gene Symbol: LOC100909840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749282.2 | XP_003749330.1 | 2 | 273 | ||

Pfn2
Genomic Map