siRNA Details
| Gene Symbol: Scn1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030875.1 | NP_110502.1 | 11 | 1819 | ||
| XM_008761923.1 | XP_008760145.1 | 11 | 1839 | |||
| XM_008761924.1 | XP_008760146.1 | 11 | 1747 | |||
| XM_008761925.1 | XP_008760147.1 | 11 | 1655 | |||
| XM_008761926.1 | XP_008760148.1 | 11 | 1838 | |||
| XM_008761927.1 | XP_008760149.1 | 11 | 1838 | |||
| XM_008761928.1 | XP_008760150.1 | 11 | 1836 | |||
| XM_008761929.1 | XP_008760151.1 | 11 | 1838 | |||
| GenBank mRNA | M22253.1 | - | 11 | 1819 | ||
| X03638.1 | - | 11 | 1819 | |||

Scn1a
Genomic Map