siRNA Details
| Gene Symbol: Acta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031004.2 | NP_112266.1 | 8, 9 | 1054 | ||
| GenBank mRNA | BC158550.1 | - | 8, 9 | 1048 | 9033322 | |
| X06801.1 | - | 8, 9 | 1039 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acta2| Gene Symbol: Acta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031004.2 | NP_112266.1 | 8, 9 | 1054 | ||
| GenBank mRNA | BC158550.1 | - | 8, 9 | 1048 | 9033322 | |
| X06801.1 | - | 8, 9 | 1039 | |||