siRNA Details
| Gene Symbol: Acta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031004.2 | NP_112266.1 | 6 | 553 | ||
| GenBank mRNA | BC158550.1 | - | 6 | 547 | 9033322 | |
| CB608976.1 | - | 2 | 148 | |||
| DN934985.1 | - | 6 | 582 | |||
| X06801.1 | - | 6 | 538 | |||

Acta2
Genomic Map