siRNA Details
| Gene Symbol: Actn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031005.3 | NP_112267.1 | 13, 14 | 1703 | ||
| GenBank mRNA | AF115386.1 | - | 13, 14 | 1703 | ||
| AY437436.1 | - | 13, 14 | 1494 | |||
| BC074001.1 | - | 1747 | 7099133 | |||

Actn1
Genomic Map