siRNA Details
| Gene Symbol: Ap2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031008.2 | NP_112270.2 | 2 | 199 | ||
| XM_008760010.1 | XP_008758232.1 | 2 | 257 | |||
| GenBank mRNA | BC081786.1 | - | 2 | 199 | 7126468 | |
| X53773.1 | - | 155 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap2a2| Gene Symbol: Ap2a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031008.2 | NP_112270.2 | 2 | 199 | ||
| XM_008760010.1 | XP_008758232.1 | 2 | 257 | |||
| GenBank mRNA | BC081786.1 | - | 2 | 199 | 7126468 | |
| X53773.1 | - | 155 | ||||