siRNA Details
| Gene Symbol: Agtr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031009.2 | NP_112271.2 | 2 | 1152 | ||
| XM_006232186.2 | XP_006232248.1 | 2 | 2322 | |||
| XM_006232187.2 | XP_006232249.1 | 2 | 2236 | |||
| XM_008760878.1 | XP_008759100.1 | 2 | 2896 | |||
| XM_008760879.1 | XP_008759101.1 | 3 | 1334 | |||
| GenBank mRNA | M90065.1 | - | 2 | 1183 | ||
| X64052.1 | - | 2 | 1140 | |||
More Information
Set Membership: |
|

Agtr1b
Genomic Map