siRNA Details
| Gene Symbol: Alox15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031010.2 | NP_112272.2 | 5 | 566 | ||
| XM_006246823.2 | XP_006246885.1 | 7 | 847 | |||
| XM_006246824.2 | XP_006246886.1 | 6 | 742 | |||
| GenBank mRNA | L06040.1 | - | 5 | 579 | ||
| S69383.1 | - | 5 | 566 | |||

Alox15
Genomic Map