siRNA Details
| Gene Symbol: Alox15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031010.2 | NP_112272.2 | 10, 11 | 1420 | ||
| XM_006246823.2 | XP_006246885.1 | 12, 13 | 1701 | |||
| XM_006246824.2 | XP_006246886.1 | 11, 12 | 1596 | |||
| GenBank mRNA | L06040.1 | - | 10, 11 | 1433 | ||
| S69383.1 | - | 10, 11 | 1420 | |||

Alox15
Genomic Map