siRNA Details
| Gene Symbol: Abcc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031013.1 | NP_112275.1 | 13 | 1791 | ||
| XM_006229159.2 | XP_006229221.1 | 13 | 1708 | |||
| XM_008759419.1 | XP_008757641.1 | 12 | 2037 | |||
| GenBank mRNA | AB010466.1 | - | 13 | 1744 | ||
| U73038.1 | - | 13 | 1791 | |||

Abcc6
Genomic Map