siRNA Details
| Gene Symbol: Opn1sw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031015.1 | NP_112277.1 | 3 | 645 | ||
| GenBank mRNA | AF051163.1 | - | 3 | 597 | ||
| U63972.1 | - | 3 | 645 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Opn1sw| Gene Symbol: Opn1sw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031015.1 | NP_112277.1 | 3 | 645 | ||
| GenBank mRNA | AF051163.1 | - | 3 | 597 | ||
| U63972.1 | - | 3 | 645 | |||
Set Membership: |
|