siRNA Details
| Gene Symbol: Atf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031018.1 | NP_112280.1 | 7, 8 | 723 | ||
| GenBank mRNA | U38938.1 | - | 7, 8 | 723 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atf2| Gene Symbol: Atf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031018.1 | NP_112280.1 | 7, 8 | 723 | ||
| GenBank mRNA | U38938.1 | - | 7, 8 | 723 | ||