siRNA Details
| Gene Symbol: Atf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031018.1 | NP_112280.1 | 12 | 1359 | ||
| GenBank mRNA | AY724488.1 | - | 937 | |||
| M65148.1 | - | 7 | 754 | |||
| U38938.1 | - | 12 | 1359 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atf2| Gene Symbol: Atf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031018.1 | NP_112280.1 | 12 | 1359 | ||
| GenBank mRNA | AY724488.1 | - | 937 | |||
| M65148.1 | - | 7 | 754 | |||
| U38938.1 | - | 12 | 1359 | |||