siRNA Details
| Gene Symbol: Crh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031019.1 | NP_112281.1 | 2 | 334 | ||
| GenBank mRNA | X03036.1 | - | 2 | 332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crh| Gene Symbol: Crh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031019.1 | NP_112281.1 | 2 | 334 | ||
| GenBank mRNA | X03036.1 | - | 2 | 332 | ||