siRNA Details
| Gene Symbol: Mapk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031020.2 | NP_112282.2 | 6 | 773 | ||
| XM_006256142.2 | XP_006256204.1 | 6 | 485 | |||
| GenBank mRNA | AB000783.1 | - | 6 | 772 | ||
| AF346293.1 | - | 6 | 470 | |||
| BC092193.1 | - | 6 | 773 | 7313217 | ||
| U73142.1 | - | 6 | 481 | |||
| U91847.1 | - | 6 | 470 | |||

Mapk14
Genomic Map