siRNA Details
| Gene Symbol: Dlat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031025.1 | NP_112287.1 | 5, 6 | 770 | ||
| GenBank mRNA | BC107440.1 | - | 5, 6 | 770 | 7115402 | |
| D00092.1 | - | 2, 3 | 274 | |||
| D10655.1 | - | 4, 5 | 487 | |||
| M16075.1 | - | 2, 3 | 274 | |||

Dlat
Genomic Map