siRNA Details
| Gene Symbol: Dlat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031025.1 | NP_112287.1 | 6 | 854 | ||
| GenBank mRNA | BC107440.1 | - | 6 | 854 | 7115402 | |
| D00092.1 | - | 3 | 358 | |||
| D10655.1 | - | 5 | 571 | |||
| M16075.1 | - | 3 | 358 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dlat| Gene Symbol: Dlat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031025.1 | NP_112287.1 | 6 | 854 | ||
| GenBank mRNA | BC107440.1 | - | 6 | 854 | 7115402 | |
| D00092.1 | - | 3 | 358 | |||
| D10655.1 | - | 5 | 571 | |||
| M16075.1 | - | 3 | 358 | |||