siRNA Details
| Gene Symbol: Gatm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031031.2 | NP_112293.1 | 3 | 506 | ||
| GenBank mRNA | AY625271.1 | - | 1 | 94 | ||
| BC081785.1 | - | 3 | 506 | 7132560 | ||
| U07971.1 | - | 3 | 498 | |||
 Back To Top
 Back To TopPlease enter the information below and press OK to send your cart to Core Services for purchase.

 Gatm
Gatm| Gene Symbol: Gatm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031031.2 | NP_112293.1 | 3 | 506 | ||
| GenBank mRNA | AY625271.1 | - | 1 | 94 | ||
| BC081785.1 | - | 3 | 506 | 7132560 | ||
| U07971.1 | - | 3 | 498 | |||
 Back To Top
 Back To Top