siRNA Details
| Gene Symbol: Gna11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031033.1 | NP_112295.1 | 4 | 505 | ||
| GenBank mRNA | AF239674.1 | - | 4 | 505 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gna11| Gene Symbol: Gna11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031033.1 | NP_112295.1 | 4 | 505 | ||
| GenBank mRNA | AF239674.1 | - | 4 | 505 | ||