siRNA Details
| Gene Symbol: Gna12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031034.2 | NP_112296.1 | 3 | 582 | ||
| GenBank mRNA | D85760.1 | - | 3 | 616 | ||
| DQ120479.1 | - | 3 | 551 | |||
| DQ120480.1 | - | 3 | 551 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gna12| Gene Symbol: Gna12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031034.2 | NP_112296.1 | 3 | 582 | ||
| GenBank mRNA | D85760.1 | - | 3 | 616 | ||
| DQ120479.1 | - | 3 | 551 | |||
| DQ120480.1 | - | 3 | 551 | |||