siRNA Details
| Gene Symbol: Gnai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031035.3 | NP_112297.1 | 1 | 204 | ||
| XM_006243858.1 | XP_006243920.1 | 1 | 234 | |||
| GenBank mRNA | AY899210.1 | - | 1 | 52 | ||
| AY899211.1 | - | 1 | 55 | |||
| BC078806.1 | - | 1 | 156 | 7121183 | ||
| CO394955.1 | - | 1 | 204 | |||
| DQ120471.1 | - | 1 | 113 | |||
| DQ120472.1 | - | 1 | 113 | |||
| M12672.1 | - | 1 | 100 | |||
| M17528.1 | - | 1 | 202 | |||

Gnai2
Genomic Map