siRNA Details
| Gene Symbol: Gpt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031039.1 | NP_112301.1 | 5 | 824 | ||
| XM_008765599.1 | XP_008763821.1 | 6 | 1544 | |||
| GenBank mRNA | BC085728.1 | - | 3 | 1050 | ||
| BC097937.1 | - | 5 | 828 | 7324977 | ||
| D10354.1 | - | 5 | 824 | |||

Gpt
Genomic Map