siRNA Details
Gene Symbol: Hnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031044.3 | NP_112306.1 | 1 | 54 | ||
GenBank mRNA | BC087635.1 | - | 1 | 245 | 7319794 | |
D10693.1 | - | 1 | 54 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hnmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031044.3 | NP_112306.1 | 1 | 54 | ||
GenBank mRNA | BC087635.1 | - | 1 | 245 | 7319794 | |
D10693.1 | - | 1 | 54 |