siRNA Details
Gene Symbol: Lss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031049.1 | NP_112311.1 | 4 | 376 | ||
GenBank mRNA | D45252.1 | - | 4 | 400 | ||
U31352.1 | - | 4 | 376 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lss | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031049.1 | NP_112311.1 | 4 | 376 | ||
GenBank mRNA | D45252.1 | - | 4 | 400 | ||
U31352.1 | - | 4 | 376 |