siRNA Details
| Gene Symbol: Mipep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031052.1 | NP_112314.1 | 10, 11 | 1182 | ||
| XM_008770779.1 | XP_008769001.1 | 10, 11 | 1335 | |||
| XM_008770780.1 | XP_008769002.1 | 10, 11 | 1335 | |||
| GenBank mRNA | M96633.1 | - | 10, 11 | 1182 | ||

Mipep
Genomic Map