siRNA Details
| Gene Symbol: Mmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031054.2 | NP_112316.2 | 11, 12 | 2071 | ||
| GenBank mRNA | BC074013.1 | - | 11, 12 | 2001 | 7100171 | |
| U65656.1 | - | 11, 12 | 2071 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mmp2| Gene Symbol: Mmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031054.2 | NP_112316.2 | 11, 12 | 2071 | ||
| GenBank mRNA | BC074013.1 | - | 11, 12 | 2001 | 7100171 | |
| U65656.1 | - | 11, 12 | 2071 | |||