siRNA Details
| Gene Symbol: Retnla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053333.1 | NP_445785.1 | 1 | 114 | ||
| XM_006248283.2 | XP_006248345.1 | 2 | 206 | |||
| GenBank mRNA | AF323085.1 | - | 1 | 114 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Retnla| Gene Symbol: Retnla | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053333.1 | NP_445785.1 | 1 | 114 | ||
| XM_006248283.2 | XP_006248345.1 | 2 | 206 | |||
| GenBank mRNA | AF323085.1 | - | 1 | 114 | ||