siRNA Details
Gene Symbol: Retnla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053333.1 | NP_445785.1 | 3 | 321 | ||
XM_006248283.2 | XP_006248345.1 | 4 | 413 | |||
GenBank mRNA | AF323085.1 | - | 3 | 321 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Retnla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053333.1 | NP_445785.1 | 3 | 321 | ||
XM_006248283.2 | XP_006248345.1 | 4 | 413 | |||
GenBank mRNA | AF323085.1 | - | 3 | 321 |