siRNA Details
Gene Symbol: Ctbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053335.1 | NP_445787.1 | 1 | 615 | ||
GenBank mRNA | AF222712.1 | - | 1 | 615 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ctbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053335.1 | NP_445787.1 | 1 | 615 | ||
GenBank mRNA | AF222712.1 | - | 1 | 615 |