siRNA Details
Gene Symbol: Mvk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031063.1 | NP_112325.1 | 7 | 883 | ||
GenBank mRNA | M29472.1 | - | 7 | 883 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mvk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031063.1 | NP_112325.1 | 7 | 883 | ||
GenBank mRNA | M29472.1 | - | 7 | 883 |