siRNA Details
| Gene Symbol: Fez1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031066.1 | NP_112328.1 | 3 | 524 | ||
| GenBank mRNA | BC087740.1 | - | 3 | 524 | 7314065 | |
| FQ212529.1 | - | 1 | 270 | |||
| U48249.2 | - | 3 | 487 | |||
| U63740.1 | - | 3 | 524 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fez1| Gene Symbol: Fez1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031066.1 | NP_112328.1 | 3 | 524 | ||
| GenBank mRNA | BC087740.1 | - | 3 | 524 | 7314065 | |
| FQ212529.1 | - | 1 | 270 | |||
| U48249.2 | - | 3 | 487 | |||
| U63740.1 | - | 3 | 524 | |||