siRNA Details
| Gene Symbol: Neo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766432.1 | XP_008764654.1 | 5 | 1021 | ||
| GenBank mRNA | U68726.1 | - | 979 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Neo1| Gene Symbol: Neo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008766432.1 | XP_008764654.1 | 5 | 1021 | ||
| GenBank mRNA | U68726.1 | - | 979 | |||