siRNA Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276711.1 | NP_001263640.1 | 24, 25 | 3113 | ||
| XM_006233358.2 | XP_006233420.1 | 22, 23 | 2769 | |||
| GenBank mRNA | FQ232433.1 | - | 7, 8 | 851 | ||
| L26267.1 | - | 10, 11 | 1409 | |||

Nfkb1
Genomic Map