siRNA Details
Gene Symbol: Nfkb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276711.1 | NP_001263640.1 | 12 | 1303 | ||
XM_006233358.2 | XP_006233420.1 | 10 | 959 | |||
XM_006233360.2 | XP_006233422.1 | 10 | 959 |
