siRNA Details
| Gene Symbol: Pcm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031076.1 | NP_112338.1 | 15 | 2594 | ||
| XM_006253183.2 | XP_006253245.1 | 17 | 3091 | |||
| XM_006253184.2 | XP_006253246.1 | 18 | 3195 | |||
| XM_006253185.2 | XP_006253247.1 | 17 | 3091 | |||
| XM_006253186.2 | XP_006253248.1 | 17 | 3090 | |||
| XM_006253187.2 | XP_006253249.1 | 16 | 2916 | |||
| XM_006253188.2 | XP_006253250.1 | 16 | 2901 | |||
| XM_008771284.1 | XP_008769506.1 | 18 | 3210 | |||
| XM_008771285.1 | XP_008769507.1 | 17 | 2939 | |||
| XM_008771286.1 | XP_008769508.1 | 17 | 2869 | |||

Pcm1
Genomic Map