siRNA Details
| Gene Symbol: Pde1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031078.2 | NP_112340.1 | 11 | 1518 | ||
| XM_008762922.1 | XP_008761144.1 | 8 | 1021 | |||
| XM_008762923.1 | XP_008761145.1 | 7 | 749 | |||
| XM_008762924.1 | XP_008761146.1 | 7 | 749 | |||
| GenBank mRNA | L41045.1 | - | 11 | 1518 | ||

Pde1c
Genomic Map