siRNA Details
| Gene Symbol: Pls3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031084.1 | NP_112346.1 | 10 | 1208 | ||
| XM_006257434.2 | XP_006257496.1 | 11 | 1343 | |||
| XM_006257438.2 | XP_006257500.1 | 10 | 1881 | |||
| XM_008773445.1 | XP_008771667.1 | 10 | 2060 | |||
| GenBank mRNA | BC092611.1 | - | 6 | 675 | ||
| X70706.1 | - | 10 | 1192 | |||

Pls3
Genomic Map